Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp4b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007823.2 | NP_031849.1 | 2 | 229 | ||
GenBank mRNA | AK143890.1 | - | 2 | 242 | ||
AK165445.1 | - | 2 | 245 | |||
BC008996.1 | - | 2 | 233 | 3967680 | ||
D50834.1 | - | 2 | 229 |