Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp8b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010012.3 | NP_034142.3 | 1 | 794 | ||
GenBank mRNA | AF090317.1 | - | 1 | 780 | ||
BC010973.1 | - | 1 | 804 | 4161038 | ||
BC010974.1 | - | 1 | 797 | 4162964 | ||
BC049969.1 | - | 1 | 779 | 4158774 |