Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dgka | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016811.2 | NP_058091.2 | 14 | 1458 | ||
GenBank mRNA | AF085219.1 | - | 13 | 1144 | ||
AK008063.1 | - | 2 | 122 | |||
AK008314.1 | - | 3 | 129 | |||
AK037536.1 | - | 1 | 1135 | |||
BC006713.1 | - | 14 | 1458 | 3584592 | ||
JN832677.1 | - | 14 | 1401 | |||
JN832678.1 | - | 14 | 1506 | |||
JN832679.1 | - | 14 | 1477 |