Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145885.1 | NP_001139357.1 | 2 | 101 | ||
NM_010029.2 | NP_034159.1 | 2 | 101 | |||
GenBank mRNA | AK014844.1 | - | 2 | 92 | ||
AK133366.1 | - | 2 | 94 | |||
BE860399.1 | - | 2 | 101 |