Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dffa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025296.2 | NP_001020467.1 | 2 | 352 | ||
NM_010044.3 | NP_034174.2 | 2 | 352 | |||
GenBank mRNA | AB009375.1 | - | 2 | 243 | ||
AB009376.1 | - | 2 | 243 | |||
AK039227.1 | - | 2 | 349 | |||
AK051011.1 | - | 2 | 282 | |||
AK079634.1 | - | 2 | 310 | |||
BC058213.1 | - | 2 | 297 | 5367607 | ||
BI736905.1 | - | 2 | 296 | |||
BY208552.1 | - | 2 | 327 | |||
BY244454.1 | - | 2 | 341 |