Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dlg4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109752.1 | NP_001103222.1 | 5, 6 | 651 | ||
NM_007864.3 | NP_031890.1 | 5, 6 | 660 | |||
GenBank mRNA | BC014807.1 | - | 5, 6 | 396 | 4501403 | |
BI738861.1 | - | 5, 6 | 405 | |||
CA320223.1 | - | 5, 6 | 651 | |||
D50621.1 | - | 5, 6 | 402 |