Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dync1h1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030238.2 | NP_084514.2 | 19 | 4339 | ||
GenBank mRNA | AK171549.1 | - | 4 | 430 | ||
AY004877.1 | - | 19 | 4293 |