Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnpep | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110831.1 | NP_001104301.1 | 14, 15 | 1684 | ||
NM_016878.4 | NP_058574.3 | 14, 15 | 1508 | |||
GenBank mRNA | AF005051.1 | - | 13, 14 | 1618 | ||
AK038646.1 | - | 1684 | ||||
AK052001.1 | - | 2075 | ||||
AK160084.1 | - | 14, 15 | 1445 | |||
AK166276.1 | - | 14, 15 | 1458 | |||
BC092232.1 | - | 14, 15 | 1436 | 30551654 |