Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnpep | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110831.1 | NP_001104301.1 | 3 | 467 | ||
NM_016878.4 | NP_058574.3 | 3 | 291 | |||
GenBank mRNA | AK038646.1 | - | 467 | |||
AK052001.1 | - | 269 | ||||
AK160084.1 | - | 3 | 228 | |||
AK166276.1 | - | 3 | 241 | |||
BC092232.1 | - | 3 | 219 | 30551654 | ||
BY197391.1 | - | 3 | 292 |