Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dpep1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007876.2 | NP_031902.2 | 6 | 800 | ||
GenBank mRNA | AK002380.1 | - | 5 | 680 | ||
BC003492.1 | - | 6 | 670 | 2812088 | ||
D13139.1 | - | 6 | 800 |