Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Drp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010078.3 | NP_034208.2 | 16 | 2174 | ||
GenBank mRNA | AK081426.1 | - | 16 | 2118 | ||
AK158102.1 | - | 15 | 1976 | |||
BC060605.1 | - | 14 | 2753 | |||
BC125345.1 | - | 16 | 2034 | 40129860 | ||
BC125347.1 | - | 16 | 2034 | 40129862 | ||
U43520.1 | - | 6 | 780 |