Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dsc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291804.1 | NP_001278733.1 | 11 | 1757 | ||
NM_013504.5 | NP_038532.3 | 11 | 1757 | |||
GenBank mRNA | AK036893.1 | - | 11 | 1643 | ||
AK079441.1 | - | 11 | 1759 | |||
AK081826.1 | - | 11 | 1665 | |||
BC109161.1 | - | 11 | 1594 | 40047883 | ||
BC109162.1 | - | 11 | 1594 | |||
X97986.1 | - | 11 | 1691 |