Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epyc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007884.2 | NP_031910.1 | 5 | 551 | ||
GenBank mRNA | AK005684.1 | - | 5 | 552 | ||
AK015223.1 | - | 5 | 551 | |||
AK137179.1 | - | 4 | 476 | |||
AK157901.1 | - | 4 | 813 | |||
BC012695.1 | - | 4 | 523 | 4037028 | ||
D78274.1 | - | 4 | 466 |