Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dvl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK155349.1 | - | 4 | 709 | ||
AK220168.1 | - | 3 | 897 | |||
BC051991.1 | - | 4 | 468 | |||
BC088987.1 | - | 4 | 573 | |||
BC138848.1 | - | 5 | 580 | 8861870 | ||
BC138849.1 | - | 5 | 580 | 8861871 | ||
BU702464.1 | - | 4 | 370 | |||
U10115.1 | - | 4 | 666 |