Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dvl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007889.2 | NP_031915.2 | 14 | 1736 | ||
GenBank mRNA | AK052151.1 | - | 14 | 1737 | ||
U41285.1 | - | 14 | 1818 |