Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ect2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177625.1 | NP_001171096.1 | 9 | 988 | ||
NM_001177626.1 | NP_001171097.1 | 9 | 1026 | |||
NM_007900.3 | NP_031926.2 | 10 | 1081 | |||
GenBank mRNA | AK040174.1 | - | 9 | 961 | ||
AK157718.1 | - | 9 | 980 | |||
AK220452.1 | - | 9 | 987 | |||
BC023881.1 | - | 9 | 986 | 5354733 | ||
BC025565.1 | - | 10 | 1055 | 5322744 | ||
BC032155.1 | - | 9 | 983 | 5067358 | ||
BC045614.1 | - | 9 | 961 | 5059799 | ||
L11316.1 | - | 9 | 926 |