Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042623.2 | NP_001036088.1 | 2 | 186 | ||
NM_001271579.1 | NP_001258508.1 | 2 | 157 | |||
NM_007905.3 | NP_031931.2 | 2 | 186 | |||
GenBank mRNA | AK036370.1 | - | 2 | 146 | ||
AK132756.1 | - | 2 | 144 | |||
AK167590.1 | - | 2 | 175 | |||
BC046535.1 | - | 2 | 167 | 4935713 | ||
BC052394.1 | - | 2 | 519 | 5719837 | ||
BU610858.1 | - | 2 | 157 | |||
CJ130893.1 | - | 2 | 186 | |||
S73882.1 | - | 2 | 142 | |||
S73883.1 | - | 2 | 142 | |||
S73884.1 | - | 2 | 142 | |||
U63386.1 | - | 1 | 108 |