Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Egf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010113.3 | NP_034243.2 | 7, 8 | 1560 | ||
GenBank mRNA | AK085707.1 | - | 7, 8 | 1127 | ||
AK088408.1 | - | 2, 3 | 147 | |||
AK136439.1 | - | 2, 3 | 153 | |||
AK142666.1 | - | 7, 8 | 1256 | |||
AK157778.1 | - | 2, 3 | 152 | |||
BC017681.1 | - | 8, 9 | 1497 | 4221592 | ||
BC060741.1 | - | 7, 8 | 1545 | 4164289 | ||
BC092277.1 | - | 7, 8 | 1545 | 4240429 | ||
J00380.1 | - | 7, 8 | 1555 | |||
V00741.1 | - | 7, 8 | 1511 |