Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Egf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010113.3 | NP_034243.2 | 23 | 3731 | ||
GenBank mRNA | AK085707.1 | - | 23 | 3298 | ||
AK088408.1 | - | 17 | 2195 | |||
AK136439.1 | - | 18 | 2324 | |||
AK142666.1 | - | 23 | 3428 | |||
BC017681.1 | - | 24 | 3668 | 4221592 | ||
BC060741.1 | - | 23 | 3716 | 4164289 | ||
BC092277.1 | - | 23 | 3716 | 4240429 | ||
J00380.1 | - | 23 | 3726 | |||
V00741.1 | - | 23 | 3682 |