Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif2ak3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010121.2 | NP_034251.2 | 15 | 3235 | ||
GenBank mRNA | AF076681.1 | - | 15 | 3227 | ||
AK003226.1 | - | 3 | 731 | |||
AK194961.1 | - | 3 | 338 | |||
BC054809.1 | - | 15 | 3247 | 6839221 | ||
CA316032.1 | - | 2 | 82 |