Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif2b4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127355.1 | NP_001120827.1 | 12 | 1552 | ||
NM_001127356.1 | NP_001120828.1 | 11 | 1774 | |||
NM_010122.2 | NP_034252.2 | 12 | 1407 | |||
GenBank mRNA | AK154364.1 | - | 12 | 1315 | ||
AK158282.1 | - | 12 | 1316 | |||
BC021884.1 | - | 12 | 1294 | 4168186 | ||
CD768292.1 | - | 3 | 302 | |||
M98035.1 | - | 11 | 1394 | |||
M98036.1 | - | 12 | 1360 |