Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007922.5 | NP_031948.4 | 2 | 295 | ||
GenBank mRNA | AK082260.1 | - | 2 | 295 | ||
AK132354.1 | - | 2 | 288 | |||
BC054474.1 | - | 2 | 279 | 30045506 | ||
CV561936.1 | - | 1 | 241 | |||
D26066.1 | - | 1 | 62 | |||
FN434115.1 | - | 1 | 29 | |||
X87257.1 | - | 2 | 565 | |||
Z36939.1 | - | 1 | 80 |