Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Emd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007927.3 | NP_031953.1 | 4, 5 | 668 | ||
GenBank mRNA | AK146361.1 | - | 4, 5 | 514 | ||
AK160531.1 | - | 4, 5 | 513 | |||
AK167852.1 | - | 4, 5 | 511 | |||
BC132131.1 | - | 4, 5 | 480 | 40130408 | ||
BC132133.1 | - | 4, 5 | 480 | 40130410 | ||
U73902.1 | - | 4, 5 | 518 |