Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Enpep | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007934.3 | NP_031960.1 | 4 | 1205 | ||
GenBank mRNA | AK031439.1 | - | 3 | 237 | ||
AK034613.1 | - | 4 | 1177 | |||
AK128975.1 | - | 4 | 1162 | |||
AK146808.1 | - | 4 | 1177 | |||
BC127060.1 | - | 4 | 1069 | 40109863 | ||
DQ000292.1 | - | 4 | 945 | |||
M29961.1 | - | 4 | 1024 |