Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Enpep | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007934.3 | NP_031960.1 | 18 | 2793 | ||
GenBank mRNA | AK031439.1 | - | 17 | 1825 | ||
AK128975.1 | - | 18 | 2750 | |||
AK146808.1 | - | 18 | 2765 | |||
BC064061.1 | - | 3 | 526 | |||
BC127060.1 | - | 18 | 2657 | 40109863 | ||
DQ000292.1 | - | 18 | 2533 | |||
M29961.1 | - | 18 | 2612 |