Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epas1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010137.3 | NP_034267.3 | 8, 9 | 1451 | ||
GenBank mRNA | AF045160.1 | - | 8, 9 | 1215 | ||
AK136911.1 | - | 8, 9 | 1455 | |||
AK141185.1 | - | 8, 9 | 1186 | |||
AK158430.1 | - | 8, 9 | 1615 | |||
BC057870.1 | - | 8, 9 | 1173 | 5032291 | ||
D89787.1 | - | 8, 9 | 1413 | |||
U81983.1 | - | 8, 9 | 1134 |