Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epas1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010137.3 | NP_034267.3 | 9 | 1651 | ||
GenBank mRNA | AF045160.1 | - | 9 | 1415 | ||
AK136911.1 | - | 9 | 1655 | |||
AK141185.1 | - | 9 | 1386 | |||
AK158430.1 | - | 9 | 1815 | |||
BC057870.1 | - | 9 | 1373 | 5032291 | ||
D89787.1 | - | 9 | 1613 | |||
U81983.1 | - | 9 | 1334 |