Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epb4.1l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199265.1 | NP_001186194.1 | 8 | 1386 | ||
NM_013511.3 | NP_038539.2 | 8 | 1277 | |||
GenBank mRNA | AB370258.1 | - | 7 | 1188 | ||
AB370259.1 | - | 7 | 1188 | |||
AB370260.1 | - | 7 | 1188 | |||
AB370263.1 | - | 7 | 1188 | |||
AF044312.1 | - | 7 | 1188 | |||
AJ428901.1 | - | 5 | 473 | |||
AJ539142.1 | - | 7 | 1188 | |||
AJ539143.1 | - | 7 | 1188 | |||
AJ539144.1 | - | 7 | 1188 | |||
AJ542537.1 | - | 7 | 1188 | |||
AK043148.1 | - | 7 | 1165 | |||
AK052475.1 | - | 8 | 1278 | |||
AK144088.1 | - | 8 | 1387 | |||
AK167434.1 | - | 8 | 1303 | |||
AK170275.1 | - | 8 | 1329 | |||
BC023220.1 | - | 8 | 1273 | |||
BC023768.1 | - | 8 | 1299 | |||
BC026708.1 | - | 8 | 1258 | |||
BC026714.1 | - | 7 | 870 | |||
BC029938.1 | - | 8 | 1415 | |||
KF534728.1 | - | 7 | 1188 |