Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epb4.1l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013813.1 | NP_038841.1 | 12, 13 | 1694 | ||
GenBank mRNA | AB370264.1 | - | 11, 12 | 1582 | ||
AB523362.1 | - | 11, 12 | 1528 | |||
AB523363.1 | - | 11, 12 | 1582 | |||
AF152247.1 | - | 12, 13 | 1694 | |||
AF177146.1 | - | 11, 12 | 1515 | |||
AK031156.1 | - | 12, 13 | 1726 | |||
AK053532.1 | - | 12, 13 | 1695 | |||
BC079858.1 | - | 12, 13 | 1758 | |||
BC152548.1 | - | 12, 13 | 1659 | 40109749 | ||
HG738869.1 | - | 5, 6 | 693 | |||
HG738870.1 | - | 11, 12 | 1582 |