Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epb4.1l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013813.1 | NP_038841.1 | 9, 10 | 1199 | ||
GenBank mRNA | AB370264.1 | - | 8, 9 | 1087 | ||
AB523362.1 | - | 8, 9 | 1087 | |||
AB523363.1 | - | 8, 9 | 1087 | |||
AF152247.1 | - | 9, 10 | 1199 | |||
AF177146.1 | - | 8, 9 | 1074 | |||
AK031156.1 | - | 9, 10 | 1231 | |||
AK053532.1 | - | 9, 10 | 1200 | |||
AK173080.1 | - | 4, 5 | 486 | |||
BC079858.1 | - | 9, 10 | 1263 | |||
BC152548.1 | - | 9, 10 | 1164 | 40109749 | ||
HG738869.1 | - | 2, 3 | 198 | |||
HG738870.1 | - | 8, 9 | 1087 |