Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dmtn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252663.1 | NP_001239592.1 | 12, 14 | 1439 | ||
NM_001252665.1 | NP_001239594.1 | 11, 13 | 1165 | |||
NM_001252666.1 | NP_001239595.1 | 10, 12 | 1198 | |||
NM_013514.4 | NP_038542.1 | 13, 15 | 1380 | |||
GenBank mRNA | AF079846.3 | - | 11, 13 | 1109 | ||
AK155927.1 | - | 12, 14 | 1434 | |||
AK162866.1 | - | 11, 13 | 1117 | |||
AK165294.1 | - | 12, 14 | 1431 | |||
BC016897.1 | - | 13, 15 | 1380 | 4240747 | ||
BC037021.1 | - | 10, 12 | 1185 | 5366040 |