Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epha1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023580.4 | NP_076069.2 | 12 | 2012 | ||
GenBank mRNA | AF131197.1 | - | 12 | 1954 | ||
AK028478.1 | - | 12 | 2013 | |||
BC071215.1 | - | 12 | 1976 | 4196138 | ||
U18084.1 | - | 8 | 1115 |