Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epha5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007937.3 | NP_031963.2 | 5, 6 | 2081 | ||
GenBank mRNA | AK089130.1 | - | 6, 7 | 1635 | ||
AK089143.1 | - | 7, 8 | 2432 | |||
BU707304.1 | - | 1, 2 | 101 | |||
U07357.1 | - | 5, 6 | 1621 |