Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ephb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010143.1 | NP_034273.1 | 11 | 2402 | ||
GenBank mRNA | AK144414.1 | - | 11 | 2419 | ||
AK158043.1 | - | 12 | 3342 | |||
BC014822.1 | - | 11 | 2403 | 3673003 | ||
BC027805.1 | - | 7 | 1689 | |||
BC053085.1 | - | 11 | 2402 | 5708827 | ||
Z49086.1 | - | 11 | 2260 |