Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ephb6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146351.1 | NP_001139823.1 | 6 | 1544 | ||
NM_007680.4 | NP_031706.3 | 6 | 1540 | |||
GenBank mRNA | AK076442.1 | - | 6 | 1366 | ||
AK087423.1 | - | 6 | 1455 | |||
BC031924.1 | - | 5 | 1152 | 4911959 | ||
L77867.1 | - | 6 | 1234 |