Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252454.1 | NP_001239383.1 | 3 | 536 | ||
NM_010147.4 | NP_034277.1 | 4 | 707 | |||
GenBank mRNA | AK155033.1 | - | 4 | 702 | ||
AK207751.1 | - | 1 | 33 | |||
BC046962.1 | - | 2 | 342 | 6391968 | ||
BC067206.1 | - | 3 | 478 | 6514327 | ||
BC099682.1 | - | 4 | 637 | 6529672 | ||
CA750240.1 | - | 4 | 708 |
Gene Symbol: Tpx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK190284.1 | - | 343 |