Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252454.1 | NP_001239383.1 | 2 | 481 | ||
NM_010147.4 | NP_034277.1 | 3 | 652 | |||
GenBank mRNA | AK155033.1 | - | 3 | 647 | ||
BC046962.1 | - | 1 | 287 | 6391968 | ||
BC067206.1 | - | 2 | 423 | 6514327 | ||
BC099682.1 | - | 3 | 582 | 6529672 | ||
CA750240.1 | - | 3 | 653 |