Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252454.1 | NP_001239383.1 | 5 | 930 | ||
NM_010147.4 | NP_034277.1 | 6 | 1101 | |||
GenBank mRNA | AK155033.1 | - | 6 | 1096 | ||
BC046962.1 | - | 4 | 736 | 6391968 | ||
BC067206.1 | - | 5 | 872 | 6514327 | ||
BC099682.1 | - | 6 | 1031 | 6529672 |