Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007946.2 | NP_031972.2 | 2 | 218 | ||
GenBank mRNA | BB843302.1 | - | 2 | 219 | ||
BC052881.1 | - | 2 | 200 | |||
D78353.1 | - | 2 | 224 | |||
L77979.1 | - | 2 | 202 |