Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007946.2 | NP_031972.2 | 5 | 705 | ||
GenBank mRNA | BC052881.1 | - | 5 | 687 | ||
D78353.1 | - | 5 | 711 | |||
L77979.1 | - | 5 | 689 |