Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Drosha | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130149.1 | NP_001123621.1 | 17 | 2578 | ||
NM_026799.3 | NP_081075.3 | 17 | 2574 | |||
GenBank mRNA | AF533013.1 | - | 13 | 1242 | ||
AK144147.1 | - | 17 | 2533 | |||
AK148640.1 | - | 17 | 2497 | |||
BC050057.1 | - | 3 | 296 | |||
BC055696.1 | - | 6 | 519 | |||
BC060265.1 | - | 14 | 1873 | |||
BC088999.1 | - | 17 | 2512 | 6417566 |