Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcl11a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242934.1 | NP_001229863.1 | 4 | 1026 | ||
NM_016707.3 | NP_057916.1 | 4 | 1026 | |||
GenBank mRNA | AF051525.1 | - | 4 | 964 | ||
AF186018.1 | - | 4 | 907 | |||
AK140949.1 | - | 4 | 971 | |||
BC010585.1 | - | 1 | 40 | 2631265 |