Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Igsf8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080419.1 | NP_536344.1 | 2 | 263 | ||
GenBank mRNA | AF411055.1 | - | 2 | 134 | ||
AF439263.1 | - | 2 | 246 | |||
AK140254.1 | - | 2 | 276 | |||
AK146104.1 | - | 2 | 276 | |||
AK146221.1 | - | 2 | 282 | |||
AK150239.1 | - | 2 | 282 | |||
AK158522.1 | - | 2 | 284 | |||
AK168363.1 | - | 2 | 282 | |||
AK206992.1 | - | 2 | 115 | |||
AK213338.1 | - | 2 | 115 | |||
BC026464.1 | - | 1 | 69 | |||
BC048387.1 | - | 2 | 263 | 6311441 | ||
BC060087.1 | - | 891 |