Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elmo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080287.2 | NP_525026.2 | 8 | 691 | ||
NM_207706.1 | NP_997589.1 | 8 | 691 | |||
GenBank mRNA | AF398884.1 | - | 7 | 590 | ||
AK032033.1 | - | 8 | 664 | |||
AK033918.1 | - | 8 | 885 | |||
AK038097.1 | - | 8 | 682 | |||
AK038455.1 | - | 8 | 797 | |||
AK045428.1 | - | 8 | 692 | |||
AK047040.1 | - | 8 | 653 | |||
AK050823.1 | - | 8 | 654 | |||
AK053574.1 | - | 8 | 655 | |||
AK129023.1 | - | 8 | 691 | |||
BC023954.1 | - | 8 | 676 | 5320429 | ||
BC033418.1 | - | 7 | 621 |