Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elmo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080287.2 | NP_525026.2 | 18 | 1843 | ||
NM_207706.1 | NP_997589.1 | 19 | 1879 | |||
GenBank mRNA | AB093301.1 | - | 9 | 2424 | ||
AF398884.1 | - | 17 | 1742 | |||
AK032033.1 | - | 19 | 1852 | |||
AK033918.1 | - | 18 | 2037 | |||
AK035710.1 | - | 3 | 229 | |||
AK038097.1 | - | 18 | 1832 | |||
AK038455.1 | - | 18 | 1949 | |||
AK045428.1 | - | 18 | 1844 | |||
AK047040.1 | - | 18 | 1805 | |||
AK050823.1 | - | 18 | 1806 | |||
AK053574.1 | - | 19 | 1843 | |||
AK129023.1 | - | 18 | 1843 | |||
AK205725.1 | - | 1 | 36 | |||
BC006678.1 | - | 4 | 446 | |||
BC023954.1 | - | 19 | 1864 | 5320429 | ||
BC033418.1 | - | 17 | 1773 |