Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acsl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK004897.1 | - | 3, 4 | 398 | ||
AK145900.1 | - | 3, 4 | 525 | |||
AK149406.1 | - | 3, 4 | 403 | |||
AK152772.1 | - | 3, 4 | 403 | |||
AK153050.1 | - | 3, 4 | 531 | |||
AK158388.1 | - | 3, 4 | 482 | |||
AK161189.1 | - | 3, 4 | 377 | |||
AK168078.1 | - | 3, 4 | 403 | |||
BC056644.1 | - | 3, 4 | 387 | 5098645 | ||
U15977.1 | - | 2, 3 | 316 |