Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007982.2 | NP_032008.2 | 30 | 3121 | ||
GenBank mRNA | AK052252.1 | - | 1 | 407 | ||
AK079821.1 | - | 32 | 3382 | |||
AK220543.1 | - | 30 | 3121 | |||
JN971016.1 | - | 31 | 2857 | |||
M95408.1 | - | 29 | 2918 |