Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130409.1 | NP_001123881.1 | 23 | 2358 | ||
NM_007982.2 | NP_032008.2 | 22 | 2203 | |||
GenBank mRNA | AB011499.1 | - | 23 | 2014 | ||
AK079821.1 | - | 23 | 2455 | |||
AK220543.1 | - | 22 | 2203 | |||
BC030180.1 | - | 23 | 2288 | |||
JN971016.1 | - | 22 | 1930 | |||
M95408.1 | - | 21 | 2000 |