Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc7a12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080852.2 | NP_543128.1 | 3 | 997 | ||
GenBank mRNA | AB055005.1 | - | 3 | 1002 |