Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007994.3 | NP_032020.2 | 3 | 440 | ||
GenBank mRNA | AJ132692.1 | - | 3 | 367 | ||
AK146946.1 | - | 3 | 493 | |||
AK166898.1 | - | 3 | 441 | |||
AK166973.1 | - | 3 | 446 | |||
AK168374.1 | - | 3 | 471 | |||
BC012720.1 | - | 3 | 445 | 4205515 | ||
D42083.1 | - | 3 | 386 |